3-Bromo-5-chloro-2-hydroxypyridine ≥97%

Supplier: Matrix Scientific

083832-1G
102840-856EA 40.25 USD
102840-856
3-Bromo-5-chloro-2-hydroxypyridine ≥97%
3-Bromo-5-chloro-2-hydroxypyridine

Hazard: Irritant

Formula: C₅H₃BrClNO
MW: 208.4400 g/mol
MDL Number: MFCD01764522
CAS Number: 137628-16-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR