2,3,4,6-Tetrachloropyridine ≥90%

Supplier: Matrix Scientific

083828-1G
102840-840EA 761.54 USD
102840-840
2,3,4,6-Tetrachloropyridine ≥90%
2,3,4,6-Tetrachloropyridine

Hazard: Irritant

Formula: C₅HCl₄N
MW: 216.88 g/mol
MDL Number: MFCD01312169
CAS Number: 14121-36-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR