5-(3-Bromophenyl)oxazole ≥97%
Supplier: Matrix Scientific
Synonyms:
5-(3-Bromophenyl)-1,3-oxazole
083811-10G
083811-1G
102840-798EA
143.08
USD
102840-798
102840-800
5-(3-Bromophenyl)oxazole ≥97%
5-(3-Bromophenyl)oxazole
Hazard: Irritant
Formula:
C₉H₆BrNO MW: 224.06 g/mol |
MDL Number:
MFCD05668970 CAS Number: 243455-57-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...