(2E)-1-(4-Bromophenyl)-3-phenylprop-2-en-1-one ≥97%

Supplier: Matrix Scientific

083796-1G
102840-752EA 467.69 USD
102840-752
(2E)-1-(4-Bromophenyl)-3-phenylprop-2-en-1-one ≥97%
(2E)-1-(4-Bromophenyl)-3-phenylprop-2-en-1-one

Hazard: Irritant

Formula: C₁₅H₁₁BrO
MDL Number: MFCD00017841
CAS Number: 2403-27-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR