2-Bromo-m-cresol ≥97%

Supplier: Matrix Scientific

Synonyms: 2-Bromo-3-methylphenol

083795-1G
102840-750EA 33.35 USD
102840-750
2-Bromo-m-cresol ≥97%
2-Bromo-m-cresol

Hazard: Irritant/Refrigerate/Stored under Argon

Formula: C₇H₇BrO
MW: 187.0300 g/mol
MDL Number: MFCD09991797
CAS Number: 22061-78-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR