3-(3-Bromophenyl)-1H-pyrazole ≥97%
Supplier: Matrix Scientific
083790-1G
102840-736EA
467.69
USD
102840-736
3-(3-Bromophenyl)-1H-pyrazole ≥97%
3-(3-Bromophenyl)-1H-pyrazole
Hazard: Irritant
Formula:
C₉H₇BrN₂ MW: 223.07 g/mol |
MDL Number:
MFCD01940433 CAS Number: 149739-65-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...