5-(4-Chlorophenyl)oxazole ≥97%

Supplier: Matrix Scientific

Synonyms: 5-(4-Chlorophenyl)-1,3-oxazole

083784-1G
102840-720EA 806.15 USD
102840-720
5-(4-Chlorophenyl)oxazole ≥97%
5-(4-Chlorophenyl)oxazole

Hazard: Irritant

Formula: C₉H₆ClNO
MW: 179.6030 g/mol
MDL Number: MFCD09258870
CAS Number: 1008-94-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR