1-(4-Iodophenyl)-1H-pyrazole ≥97%

Supplier: Matrix Scientific

083777-1G
102840-696EA 261.54 USD
102840-696
1-(4-Iodophenyl)-1H-pyrazole ≥97%
1-(4-Iodophenyl)-1H-pyrazole

Hazard: Irritant

Formula: C₉H₇IN₂
MW: 270.0700 g/mol
MDL Number: MFCD02681941
CAS Number: 368869-86-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR