4-Iodophenylsulfur pentafluoride ≥99%

Supplier: Matrix Scientific

083772-1G
102840-684EA 553.85 USD
102840-684
4-Iodophenylsulfur pentafluoride ≥99%
4-Iodophenylsulphur pentafluoride

Hazard: Irritant

Formula: C₆H₄F₅IS
MDL Number: MFCD03788514
CAS Number: 286947-68-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR