3-Bromophenylsulfur pentafluoride ≥98%

Supplier: Matrix Scientific

083771-1G
102840-682EA 553.85 USD
102840-682
3-Bromophenylsulfur pentafluoride ≥98%
3-Bromophenylsulphur pentafluoride

Hazard: Irritant

Formula: C₆H₄BrF₅S
MW: 283.06 g/mol
MDL Number: MFCD03425927
CAS Number: 672-30-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR