3-(Pyrimidin-2-yl)bromobenzene ≥97%

Supplier: Matrix Scientific

Synonyms: 2-(3-Bromophenyl)pyrimidine

083766-1G
102840-668EA 609.23 USD
102840-668
3-(Pyrimidin-2-yl)bromobenzene ≥97%
3-(Pyrimidin-2-yl)bromobenzene

Hazard: Irritant

Formula: C₁₀H₇BrN₂
MW: 235.09 g/mol
MDL Number: MFCD08056330
CAS Number: 885468-36-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR