4-(Pyrimidin-2-yl)bromobenzene ≥97%

Supplier: Matrix Scientific

Synonyms: 2-(4-Bromophenyl)pyrimidine

083765-1G
102840-666EA 638.46 USD
102840-666
4-(Pyrimidin-2-yl)bromobenzene ≥97%
4-(Pyrimidin-2-yl)bromobenzene

Hazard: Irritant

Formula: C₁₀H₇BrN₂
MW: 235.09 g/mol
MDL Number: MFCD08056329
CAS Number: 210354-17-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR