5-(4-Bromophenyl)-3-methyl-1,2,4-oxadiazole ≥97%

Supplier: Matrix Scientific

083757-1G
102840-644EA 687.69 USD
102840-644
5-(4-Bromophenyl)-3-methyl-1,2,4-oxadiazole ≥97%
5-(4-Bromophenyl)-3-methyl-1,2,4-oxadiazole

Hazard: Irritant

Formula: C₉H₇BrN₂O
MDL Number: MFCD03069782
CAS Number: 71566-07-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR