5-(2-Fluorophenyl)cyclohexane-1,3-dione ≥97%

Supplier: Matrix Scientific

083744-1G
102840-612EA 547.69 USD
102840-612
5-(2-Fluorophenyl)cyclohexane-1,3-dione ≥97%
5-(2-Fluorophenyl)cyclohexane-1,3-dione

Hazard: Irritant

Formula: C₁₂H₁₁FO₂
MDL Number: MFCD03844401
CAS Number: 102821-72-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR