Ethyl-4,4,4-trichloroacetoacetate ≥97%

Supplier: Matrix Scientific

Synonyms: 4,4,4-Trichloroacetoacetic acid ethyl ester

083729-1G 083729-10G
102840-564EA 127.69 USD
102840-564 102840-566
Ethyl-4,4,4-trichloroacetoacetate ≥97%
Ethyl 4,4,4-trichloroacetoacetate

Hazard: Irritant

Formula: C₆H₇Cl₃O₃
MDL Number: MFCD00043329
CAS Number: 3702-98-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR