tert-Butyl (1S)-1-(hydroxymethyl)-2-methylpropylcarbamate ≥97%

Supplier: Matrix Scientific

Synonyms: N-BOC-L-ValinolBoc-Valinol(S)-tert-butyl (1-hydroxy-3-methylbutan-2-yl)carbamate

083723-10G 083723-1G
102840-546EA 156.92 USD
102840-546 102840-548
tert-Butyl (1S)-1-(hydroxymethyl)-2-methylpropylcarbamate ≥97%
tert-Butyl (1S)-1-(hydroxymethyl)-2-methylpropylcarbamate

Hazard: Irritant

Formula: C₁₀H₂₁NO₃
MW: 203.28 g/mol
MDL Number: MFCD00082635
CAS Number: 79069-14-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR