tert-Butyl-4-aminobutyl(methyl)carbamate ≥97%

Supplier: Matrix Scientific

Synonyms: tert-Butyl 4-aminobutyl(methyl)carbamate

083721-1G
102840-538EA 524.62 USD
102840-538
tert-Butyl-4-aminobutyl(methyl)carbamate ≥97%
tert-Butyl-4-aminobutyl(methyl)carbamate

Hazard: Irritant

Formula: C₁₀H₂₂N₂O₂
MW: 202.2900 g/mol
MDL Number: MFCD03001681
CAS Number: 144222-23-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR