2-(Ethoxycarbonyl)-3-propylhexanoic acid ≥97%

Supplier: Matrix Scientific

083718-1G
102840-530EA 524.62 USD
102840-530
2-(Ethoxycarbonyl)-3-propylhexanoic acid ≥97%
2-(Ethoxycarbonyl)-3-propylhexanoic acid

Hazard: Irritant

Formula: C₁₂H₂₂O₄
MDL Number: MFCD00829131
CAS Number: 255714-16-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR