L-Valine ethyl ester hydrochloride ≥97%
Supplier: Matrix Scientific
Synonyms:
H-Val-OEt·HCl
, (S)-Ethyl 2-amino-3-methylbutanoate hydrochloride
083715-10G
083715-1G
083715-50G
102840-518EA
1030.77
USD
102840-518
102840-514
102840-516
L-Valine ethyl ester hydrochloride ≥97%
L-Valine ethyl ester hydrochloride
Hazard: Irritant
Formula:
C₇H₁₆ClNO₂ MW: 181.66 g/mol |
MDL Number:
MFCD00012511 CAS Number: 17609-47-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...