Dimethyl (1-ethylpropyl)malonate ≥97%
Supplier: Matrix Scientific
083708-10G
083708-1G
102840-488EA
241.54
USD
102840-488
102840-490
Dimethyl (1-ethylpropyl)malonate ≥97%
Dimethyl (1-ethylpropyl)malonate
Hazard: Irritant
Formula:
C₁₀H₁₈O₄ |
MDL Number:
MFCD08436116 CAS Number: 39520-19-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...