Ethyl (3E)-3-(acetylhydrazono)-2-chlorobutanoate ≥97%

Supplier: Matrix Scientific

083700-1G 083700-10G
102840-460EA 184.62 USD
102840-460 102840-462
Ethyl (3E)-3-(acetylhydrazono)-2-chlorobutanoate ≥97%
Ethyl (3E)-3-(acetylhydrazono)-2-chlorobutanoate

Hazard: Irritant

Formula: C₈H₁₃ClN₂O₃
MDL Number: MFCD09991794
CAS Number: 66552-43-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR