Diethyl diallylmalonate ≥97%
Supplier: Matrix Scientific
Synonyms:
Diallylmalonic Acid Diethyl Ester
, Diethyl 2,2-diallylmalonate
083693-50G
083693-10G
083693-1G
102840-434EA
72.31
USD
102840-434
102840-436
102840-438
Diethyl diallylmalonate ≥97%
Diethyl diallylmalonate
Purity: 97%
CAS: 3195-24-2
MDL Number: MFCD00009126
Molecular Formula: C13H20O4
Molecular Weight: 240,3
Hazard: Irritant
Formula:
C₁₃H₂₀O₄ MW: 240.30 g/mol |
MDL Number:
MFCD00009126 CAS Number: 3195-24-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...