D-Cysteine methyl ester hydrochloride ≥97%
Supplier: Matrix Scientific
Synonyms:
H-D-Cys-OMe.HCl
083692-1G
102840-432EA
241.54
USD
102840-432
D-Cysteine methyl ester hydrochloride ≥97%
D-Cysteine methyl ester hydrochloride
Purity: 97%
CAS: 70361-61-4
MDL Number: MFCD00173915
Molecular Formula: C4H10ClNO2S
Molecular Weight: 171,65
Hazard: Irritant
Formula:
C₄H₉NO₂S MW: 171.64 g/mol |
MDL Number:
MFCD00173915 CAS Number: 70361-61-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...