Ethyl [(4-chlorophenyl)sulfonyl]acetate ≥97%

Supplier: Matrix Scientific

083690-1G
102840-426EA 863.08 USD
102840-426
Ethyl [(4-chlorophenyl)sulfonyl]acetate ≥97%
Ethyl [(4-chlorophenyl)sulfonyl]acetate

Hazard: Irritant

Formula: C₁₀H₁₁ClO₄S
MDL Number: MFCD00018662
CAS Number: 3636-65-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR