Ethyl-4,5,6,7-tetrahydroisoindole-1-carboxylate ≥97%

Supplier: Matrix Scientific

Synonyms: 4,5,6,7-Tetrahydroisoindole-1-carboxylic Acid Ethyl Ester

083679-1G
102840-386EA 524.62 USD
102840-386
Ethyl-4,5,6,7-tetrahydroisoindole-1-carboxylate ≥97%
Ethyl-4,5,6,7-tetrahydroisoindole-1-carboxylate

Purity: 97%
CAS: 65880-17-3
MDL Number: MFCD06227773
Molecular Formula: C11H15NO2
Molecular Weight: 193,25
Hazard: Irritant

Formula: C₁₁H₁₅NO₂
MW: 193.25 g/mol
MDL Number: MFCD06227773
CAS Number: 65880-17-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR