Ethyl-1-oxo-1H-pyrano[3,4-c]pyridine-3-carboxylate ≥97%

Supplier: Matrix Scientific

083668-1G
102840-360EA 750.77 USD
102840-360
Ethyl-1-oxo-1H-pyrano[3,4-c]pyridine-3-carboxylate ≥97%
Ethyl-1-oxo-1H-pyrano[3,4-c]pyridine-3-carboxylate

Hazard: Irritant

Formula: C₁₁H₉NO₄
MDL Number: MFCD13195612
CAS Number: 58790-51-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR