Ethyl-3-bromoimidazo[1,2-a]pyridine-6-carboxylate ≥97%
Supplier: Matrix Scientific
083665-1G
102840-354EA
949.23
USD
102840-354
Ethyl-3-bromoimidazo[1,2-a]pyridine-6-carboxylate ≥97%
Ethyl 3-bromoimidazo[1,2-a]pyridine-6-carboxylate
Hazard: Irritant
Formula:
C₁₀H₉BrN₂O₂ MW: 269.10 |
MDL Number:
MFCD12827886 CAS Number: 1215504-30-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...