Ethyl-1-cyanocyclobutanecarboxylate ≥97%

Supplier: Matrix Scientific

Synonyms: Ethyl 1-cyanocyclobutanecarboxylate

083631-1G
102840-276EA 863.08 USD
102840-276
Ethyl-1-cyanocyclobutanecarboxylate ≥97%
Ethyl-1-cyanocyclobutanecarboxylate

Hazard: Irritant

Formula: C₈H₁₁NO₂
MW: 153.18 g/mol
MDL Number: MFCD04114273
CAS Number: 28246-87-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR