Ethyl-3-cyclohexene-1-carboxylate ≥97%
Supplier: Matrix Scientific
Synonyms:
3-Cyclohexene-1-carboxylic Acid Ethyl Ester
083609-1G
083609-50G
083609-10G
102840-220EA
1127.69
USD
102840-220
102840-216
102840-218
Ethyl-3-cyclohexene-1-carboxylate ≥97%
Ethyl-3-cyclohexene-1-carboxylate
Purity: 97%
CAS: 15111-56-5
MDL Number: MFCD00037158
Molecular Formula: C9H14O2
Molecular Weight: 154,21
Hazard: Irritant
Formula:
C₉H₁₄O₂ MW: 154.21 g/mol |
MDL Number:
MFCD00037158 CAS Number: 15111-56-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...