(2R)-Oxiran-2-ylmethyl pentanoate ≥97%

Supplier: Matrix Scientific

083608-1G 083608-50G 083608-10G
102840-210EA 56.92 USD
102840-210 102840-212 102840-214
(2R)-Oxiran-2-ylmethyl pentanoate ≥97%
(2R)-Oxiran-2-ylmethyl pentanoate

Hazard: Irritant

Formula: C₈H₁₄O₃
MDL Number: MFCD09991792
CAS Number: 92418-73-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR