Methyl-4-hydroxycyclohexanecarboxylate ≥97%
Supplier: Matrix Scientific
Synonyms:
Methyl 4-hydroxycyclohexanecarboxylate
083607-1G
102840-206EA
558.46
USD
102840-206
Methyl-4-hydroxycyclohexanecarboxylate ≥97%
Methyl-4-hydroxycyclohexanecarboxylate
Hazard: Irritant
Formula:
C₈H₁₄O₃ MW: 158.2 g/mol |
MDL Number:
MFCD00729087 CAS Number: 17449-76-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...