Ethyl phenyl(piperidin-2-yl)acetate hydrochloride ≥97%

083560-1G
102840-094EA 607.42 USD
102840-094
Ethyl phenyl(piperidin-2-yl)acetate hydrochloride ≥97%
Ethyl phenyl(piperidin-2-yl)acetate hydrochloride

Hazard: Irritant

Formula: C₁₅H₂₂ClNO₂
MDL Number: MFCD16622249
CAS Number: 19716-79-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR