Ethyl (1-methylpiperidin-4-yl)acetate ≥97%
Supplier: Matrix Scientific
083559-1G
102840-092EA
723.08
USD
102840-092
Ethyl (1-methylpiperidin-4-yl)acetate ≥97%
Ethyl (1-methylpiperidin-4-yl)acetate
Hazard: Irritant
Formula:
C₁₀H₁₉NO₂ |
MDL Number:
MFCD11100758 CAS Number: 67686-05-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...