3-Ethyl-2-methyl-6-oxooctahydroisoquinoline-2,3(1H)-dicarboxylate ≥97%

Supplier: Matrix Scientific

083550-1G
102840-064EA 1118.46 USD
102840-064
3-Ethyl-2-methyl-6-oxooctahydroisoquinoline-2,3(1H)-dicarboxylate ≥97%
3-Ethyl-2-methyl-6-oxooctahydroisoquinoline-2,3(1H)-dicarboxylate

Hazard: Irritant

Formula: C₁₄H₂₁NO₅
MDL Number: MFCD11505033
CAS Number: 128073-42-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR