Ethyl-6-oxo-1,6-dihydropyridazine-3-carboxylate ≥97%

Supplier: Matrix Scientific

083508-1G
102839-966EA 636.92 USD
102839-966
Ethyl-6-oxo-1,6-dihydropyridazine-3-carboxylate ≥97%
Ethyl-6-oxo-1,6-dihydropyridazine-3-carboxylate

Hazard: Irritant

Formula: C₇H₈N₂O₃
MDL Number: MFCD00160459
CAS Number: 63001-81-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR