Methyl-2-chloro-6-methylpyrimidine-4-carboxylate ≥97%

Supplier: Matrix Scientific

Synonyms: Methyl 2-chloro-6-methylpyrimidine-4-carboxylate

083505-1G
102839-960EA 213.85 USD
102839-960
Methyl-2-chloro-6-methylpyrimidine-4-carboxylate ≥97%
Methyl-2-chloro-6-methylpyrimidine-4-carboxylate

Hazard: Irritant

Formula: C₇H₇ClN₂O₂
MW: 186.6 g/mol
MDL Number: MFCD00117906
CAS Number: 89793-11-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR