Ethyl-2-chloro-4-(trifluoromethyl)pyrimidine-5-carboxylate ≥97%

Supplier: Matrix Scientific

Synonyms: Ethyl 2-chloro-4-(trifluoromethyl)pyrimidine-5-carboxylate

083501-1G 083501-10G
102839-948EA 904.62 USD
102839-948 102839-946
Ethyl-2-chloro-4-(trifluoromethyl)pyrimidine-5-carboxylate ≥97%
Ethyl-2-chloro-4-(trifluoromethyl)pyrimidine-5-carboxylate

Hazard: Irritant

Formula: C₈H₆ClF₃N₂O₂
MW: 254.6 g/mol
MDL Number: MFCD01312372
CAS Number: 187035-79-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR