Ethyl-5-oxazolecarboxylate ≥97%

Supplier: Matrix Scientific

Synonyms: Ethyl oxazole-5-carboxylate

083491-1G
102839-916EA 392.15 USD
102839-916
Ethyl-5-oxazolecarboxylate ≥97%
Ethyl 5-oxazolecarboxylate

Hazard: Irritant

Formula: C₆H₇NO₃
MW: 141.13 g/mol
MDL Number: MFCD04114930
CAS Number: 118994-89-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR