Methyl-5-methyl-4-oxazolecarboxylate ≥97%

Supplier: Matrix Scientific

Synonyms: Methyl 5-methyloxazole-4-carboxylate

083490-1G
102839-914EA 292.31 USD
102839-914
Methyl-5-methyl-4-oxazolecarboxylate ≥97%
Methyl 5-methyl-4-oxazolecarboxylate

Hazard: Irritant

Formula: C₆H₇NO₃
MW: 141.1247 g/mol
MDL Number: MFCD08689502
CAS Number: 41172-57-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR