Methyl-4-(1-hydroxyiminoethyl)-5-methylisoxazole-3-carboxylate ≥97%

Supplier: Matrix Scientific

083477-1G
102839-880EA 353.85 USD
102839-880
Methyl-4-(1-hydroxyiminoethyl)-5-methylisoxazole-3-carboxylate ≥97%
Methyl-4-(1-hydroxyiminoethyl)-5-methylisoxazole-3-carboxylate

Hazard: Irritant

Formula: C₈H₁₀N₂O₄
MW: 198.18 g/mol
MDL Number: MFCD00068117
CAS Number: 175277-20-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR