Ethyl-4-methyl-1,3-thiazole-2-carboxylate ≥97%

Supplier: Matrix Scientific

083474-1G
102839-872EA 636.92 USD
102839-872
Ethyl-4-methyl-1,3-thiazole-2-carboxylate ≥97%
Ethyl-4-methyl-1,3-thiazole-2-carboxylate

Hazard: Irritant

Formula: C₇H₉NO₂S
MW: 171.22 g/mol
Boiling Pt: 130…132 °C (16 Torr)
MDL Number: MFCD11112113
CAS Number: 7210-73-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR