Ethyl-2-(benzylamino)-1,3-thiazole-5-carboxylate ≥97%

Supplier: Matrix Scientific

083473-1G
102839-870EA 270.77 USD
102839-870
Ethyl-2-(benzylamino)-1,3-thiazole-5-carboxylate ≥97%
Ethyl-2-(benzylamino)-1,3-thiazole-5-carboxylate

Hazard: Irritant

Formula: C₁₃H₁₄N₂O₂S
MW: 262.33 g/mol
MDL Number: MFCD03086199
CAS Number: 342394-00-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR