Ethyl-3,4-dimethyl-1H-pyrrole-2-carboxylate ≥97%

Supplier: Matrix Scientific

083460-1G
102839-840EA 676.92 USD
102839-840
Ethyl-3,4-dimethyl-1H-pyrrole-2-carboxylate ≥97%
Ethyl-3,4-dimethyl-1H-pyrrole-2-carboxylate

Hazard: Irritant

Formula: C₉H₁₃NO₂
MW: 167.21 g/mol
MDL Number: MFCD00030365
CAS Number: 938-75-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR