Methyl-5-nitro-2-furoate ≥97%
Supplier: Matrix Scientific
083449-50G
083449-1G
083449-10G
102839-814EA
2030.13
USD
102839-814
102839-812
102839-810
Methyl-5-nitro-2-furoate ≥97%
Methyl 5-nitro-2-furoate
Hazard: Irritant
Formula:
C₆H₅NO₅ MW: 171.11 g/mol |
MDL Number:
MFCD00051956 CAS Number: 1874-23-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...