Methyl-3-isothiocyanato-4-methylthiophene-2-carboxylate ≥97%

Supplier: Matrix Scientific

083444-1G
102839-800EA 184.62 USD
102839-800
Methyl-3-isothiocyanato-4-methylthiophene-2-carboxylate ≥97%
Methyl-3-isothiocyanato-4-methylthiophene-2-carboxylate

Hazard: Irritant

Formula: C₈H₇NO₂S₂
MW: 213.28 g/mol
MDL Number: MFCD00052073
CAS Number: 81321-15-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR