Methyl-3-amino-4-(isopropylsulfonyl)thiophene-2-carboxylate ≥97%
Supplier: Matrix Scientific
083442-1G
102839-796EA
200
USD
102839-796
Methyl-3-amino-4-(isopropylsulfonyl)thiophene-2-carboxylate ≥97%
Methyl-3-amino-4-(isopropylsulfonyl)thiophene-2-carboxylate
Hazard: Irritant
Formula:
C₉H₁₃NO₄S₂ |
MDL Number:
MFCD00067930 CAS Number: 175201-72-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...