Methyl-4-amino-3-chloro-5-nitrobenzoate ≥97%
Supplier: Matrix Scientific
Synonyms:
Methyl 4-amino-3-chloro-5-nitrobenzoate
083350-1G
083350-10G
102839-528EA
1163.08
USD
102839-528
102839-526
Methyl-4-amino-3-chloro-5-nitrobenzoate ≥97%
Methyl 4-amino-3-chloro-5-nitrobenzoate
Hazard: Irritant
Formula:
C₈H₇ClN₂O₄ MW: 230.6052 g/mol |
MDL Number:
MFCD09842615 CAS Number: 863886-04-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...