Methyl-5-amino-2-methoxybenzoate ≥97%

Supplier: Matrix Scientific

Synonyms: Methyl 5-amino-2-methoxybenzoate

083338-1G
102839-496EA 383.08 USD
102839-496
Methyl-5-amino-2-methoxybenzoate ≥97%
Methyl-5-amino-2-methoxybenzoate

Hazard: Irritant

Formula: C₉H₁₁NO₃
MW: 181.19 g/mol
MDL Number: MFCD08458835
CAS Number: 22802-67-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR