Methyl-2-methyl-4-nitrobenzoate ≥97%
Supplier: Matrix Scientific
Synonyms:
Methyl 2-methyl-4-nitrobenzoate
083334-1G
102839-486EA
126.15
USD
102839-486
Methyl-2-methyl-4-nitrobenzoate ≥97%
Methyl 2-methyl-4-nitrobenzoate
Hazard: Irritant
Formula:
C₉H₉NO₄ MW: 195.1700 g/mol |
MDL Number:
MFCD03618407 CAS Number: 62621-09-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...