Methyl-3-aminosalicylate ≥97%
Supplier: Matrix Scientific
Synonyms:
Methyl 3-amino-2-hydroxybenzoate
083328-1G
102839-470EA
136.92
USD
102839-470
Methyl-3-aminosalicylate ≥97%
Methyl 3-aminosalicylate
Hazard: Irritant
Formula:
C₈H₉NO₃ MW: 167.17 |
MDL Number:
MFCD09842614 CAS Number: 35748-34-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...